Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278534.1 | NP_001265463.1 | 1 | 568 | ||
GenBank mRNA | AK032151.1 | - | 1 | 458 | ||
AK220436.1 | - | 1 | 565 | |||
BC132391.1 | - | 1 | 422 | 40130668 | ||
BC137795.1 | - | 1 | 422 | 8860814 |