Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198109.4 | NP_932777.2 | 6 | 440 | ||
GenBank mRNA | AB099287.1 | - | 6 | 475 | ||
AK089308.1 | - | 6 | 444 | |||
AK155557.1 | - | 6 | 465 | |||
AK165380.1 | - | 5 | 405 | |||
AK170787.1 | - | 6 | 431 | |||
AK171210.1 | - | 6 | 489 | |||
AK171556.1 | - | 6 | 455 | |||
AK173144.1 | - | 6 | 462 |