Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005763.3 | NP_005754.2 | 2 | 321 | ||
XM_011515725.2 | XP_011514027.1 | 2 | 300 | |||
GenBank mRNA | AF229180.1 | - | 2 | 245 | ||
AJ007714.2 | - | 1 | 272 | |||
AK225898.1 | - | 2 | 214 | |||
BC144247.1 | - | 1 | 231 | |||
DA995180.1 | - | 2 | 314 |