Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167828.1 | NP_001161300.1 | 3 | 460 | ||
NM_199146.2 | NP_954597.2 | 4 | 936 | |||
GenBank mRNA | AK136456.1 | - | 3 | 464 | ||
AK164938.1 | - | 3 | 459 | |||
BC056631.1 | - | 4 | 912 | 5035734 |