Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144552.3 | NP_653135.2 | 5, 6 | 941 | ||
GenBank mRNA | AK158779.1 | - | 5, 6 | 827 | ||
BC024598.1 | - | 5, 6 | 823 | 4020199 |