Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144835.4 | NP_659084.3 | 16 | 2145 | ||
GenBank mRNA | AK044969.1 | - | 16 | 2141 | ||
AK143958.1 | - | 16 | 2123 | |||
AK165368.1 | - | 16 | 2087 | |||
AK165890.1 | - | 16 | 2148 | |||
AK172508.1 | - | 16 | 2142 | |||
BC024471.1 | - | 13 | 1596 |