Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144919.2 | NP_659168.1 | 8 | 627 | ||
GenBank mRNA | AK045994.1 | - | 8 | 639 | ||
AK181630.1 | - | 4 | 176 | |||
BC016208.1 | - | 8 | 627 | 4913322 |