Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145218.3 | NP_660253.2 | 7 | 2083 | ||
GenBank mRNA | AB175684.1 | - | 7 | 1161 | ||
AF467979.1 | - | 7 | 1364 | |||
AK035817.1 | - | 7 | 1837 | |||
AK048758.1 | - | 7 | 1828 | |||
AK051281.1 | - | 7 | 2483 | |||
BC052469.1 | - | 7 | 1479 | 6810287 | ||
BC069177.1 | - | 7 | 2083 |