Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145425.3 | NP_663400.2 | 4, 5 | 285 | ||
GenBank mRNA | AF424701.1 | - | 4, 5 | 248 | ||
AK043672.1 | - | 3, 4 | 172 | |||
AK083042.1 | - | 4, 5 | 286 | |||
BC141166.1 | - | 4, 5 | 249 | 9055778 |