Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uts2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145440.1 | NP_663415.1 | 1 | 1008 | ||
GenBank mRNA | AF441863.1 | - | 1 | 1008 | ||
AY065981.1 | - | 1 | 967 | |||
BC125312.1 | - | 1 | 987 | 40129827 | ||
BC125318.1 | - | 1 | 987 | 40129833 |
Set Membership: |
G-protein coupled receptor |