Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145452.3 | NP_663427.2 | 4 | 1416 | ||
GenBank mRNA | BC013637.1 | - | 2 | 162 | 3498774 | |
BC065082.1 | - | 4 | 959 |