Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145471.2 | NP_663446.1 | 5 | 968 | ||
GenBank mRNA | AK036278.1 | - | 3 | 841 | ||
AK053747.1 | - | 1078 | ||||
AK132158.1 | - | 3 | 1532 | |||
AK149655.1 | - | 3 | 930 | |||
AK159836.1 | - | 3 | 911 | |||
AK172871.1 | - | 3 | 1507 | |||
AY239225.1 | - | 3 | 1202 | |||
AY239226.1 | - | 5 | 884 | |||
BC022126.1 | - | 5 | 1001 | 5097371 | ||
BC033482.1 | - | 2 | 935 | |||
BC138217.1 | - | 3 | 1142 | 8861236 |