Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145471.2 | NP_663446.1 | 6 | 1743 | ||
GenBank mRNA | AK036278.1 | - | 4 | 1616 | ||
AK047414.1 | - | 4 | 1288 | |||
AK053747.1 | - | 1853 | ||||
AK132158.1 | - | 4 | 2307 | |||
AK149655.1 | - | 4 | 1706 | |||
AK159836.1 | - | 4 | 1686 | |||
AK172871.1 | - | 4 | 2282 | |||
AY239225.1 | - | 4 | 1977 | |||
AY239226.1 | - | 6 | 1659 | |||
AY344668.1 | - | 4 | 1293 | |||
BC022126.1 | - | 6 | 1776 | 5097371 | ||
BC033482.1 | - | 3 | 1710 | |||
BC138217.1 | - | 4 | 1917 | 8861236 | ||
BC145203.1 | - | 4 | 1354 |