Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145476.2 | NP_663451.2 | 4, 5 | 760 | ||
GenBank mRNA | AK039627.1 | - | 4, 5 | 773 | ||
AK146872.1 | - | 4, 5 | 761 | |||
AK154226.1 | - | 4, 5 | 782 | |||
AK154602.1 | - | 4, 5 | 803 | |||
AK155459.1 | - | 4, 5 | 774 | |||
AK155770.1 | - | 4, 5 | 803 | |||
AK178650.1 | - | 2, 3 | 421 | |||
BC023106.1 | - | 1, 2 | 155 | 4016550 | ||
BC066009.1 | - | 4, 5 | 760 | 5708486 |