Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145516.2 | NP_663491.1 | 3 | 208 | ||
NM_175421.1 | NP_780630.1 | 3 | 208 | |||
GenBank mRNA | AF189817.1 | - | 3 | 168 | ||
AK039771.1 | - | 3 | 176 | |||
AK082118.1 | - | 3 | 161 | |||
AK148653.1 | - | 3 | 191 | |||
AK149868.1 | - | 3 | 168 | |||
AK150074.1 | - | 3 | 168 | |||
AK166091.1 | - | 3 | 176 | |||
AK179998.1 | - | 3 | 121 | |||
AK187330.1 | - | 121 | ||||
BC057994.1 | - | 3 | 176 | 5344583 | ||
CK625615.1 | - | 3 | 208 |