Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 4 | 418 | ||
GenBank mRNA | AK050860.1 | - | 4 | 378 | ||
AK086699.1 | - | 4 | 411 | |||
AK171713.1 | - | 4 | 444 | |||
BB663505.1 | - | 4 | 411 | |||
BC009153.1 | - | 4 | 407 | 2655209 | ||
BI739824.1 | - | 2 | 43 |