Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145548.4 | NP_663523.2 | 9 | 1699 | ||
GenBank mRNA | AK143903.1 | - | 9 | 1700 | ||
AK143981.1 | - | 9 | 1695 | |||
AK165405.1 | - | 9 | 1699 | |||
AK165469.1 | - | 9 | 1677 | |||
AK165473.1 | - | 9 | 1677 | |||
BC016446.1 | - | 7 | 1373 | 4235131 | ||
BC024451.1 | - | 7 | 1180 |