Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tardbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003898.3 | NP_001003898.1 | 4 | 808 | ||
NM_001003899.2 | NP_001003899.1 | 4 | 808 | |||
NM_001008545.2 | NP_001008545.1 | 4 | 808 | |||
NM_001008546.2 | NP_001008546.1 | 4 | 808 | |||
NM_145556.4 | NP_663531.1 | 4 | 808 | |||
GenBank mRNA | AK032199.1 | - | 4 | 578 | ||
AK045495.1 | - | 4 | 578 | |||
AK083312.1 | - | 4 | 582 | |||
AK133207.1 | - | 4 | 585 | |||
AK146002.1 | - | 4 | 578 | |||
AK146997.1 | - | 5 | 487 | |||
AK153803.1 | - | 4 | 582 | |||
AY319513.1 | - | 3 | 478 | |||
AY319514.1 | - | 3 | 478 | |||
AY319515.1 | - | 3 | 478 | |||
AY319516.1 | - | 3 | 478 | |||
BC012873.1 | - | 4 | 566 | 4016437 | ||
BC025544.1 | - | 4 | 579 | 5321007 | ||
BC027105.1 | - | 4 | 570 | 5359082 | ||
BC027772.1 | - | 4 | 568 | 5346061 | ||
BC029728.1 | - | 4 | 569 | 4950263 | ||
BC031126.1 | - | 4 | 570 | 4977114 | ||
BC033475.1 | - | 4 | 566 | 4953334 | ||
GQ428775.1 | - | 3 | 478 | |||
GQ428776.1 | - | 3 | 478 | |||
GQ428777.1 | - | 3 | 478 | |||
GU250735.1 | - | 3 | 478 |