Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145743.2 | NP_665686.2 | 1, 2 | 311 | ||
GenBank mRNA | AF520417.1 | - | 1, 2 | 160 | ||
AK044595.1 | - | 1, 2 | 187 | |||
AK170996.1 | - | 1, 2 | 162 | |||
BC089595.1 | - | 1, 2 | 161 | 30118411 | ||
BY295329.1 | - | 1, 2 | 311 |