Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145974.3 | NP_666086.2 | 5 | 2445 | ||
GenBank mRNA | AK049249.1 | - | 5 | 2215 | ||
AK141228.1 | - | 5 | 2239 | |||
AK142500.1 | - | 5 | 2208 | |||
AK170521.1 | - | 5 | 2240 | |||
AK218573.1 | - | 1 | 267 | |||
BC005767.1 | - | 2 | 992 | |||
BC006740.1 | - | 5 | 2158 | 3158173 | ||
BC055831.1 | - | 5 | 2445 | 4187601 |