Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005761.2 | NP_005752.1 | 12, 13 | 2634 | ||
XM_006719186.3 | XP_006719249.1 | 12, 13 | 2661 | |||
XM_011537730.2 | XP_011536032.1 | 12, 13 | 2661 | |||
XM_011537731.2 | XP_011536033.1 | 12, 13 | 2661 | |||
XM_017018671.1 | XP_016874160.1 | 12, 13 | 2661 | |||
XM_017018672.1 | XP_016874161.1 | 12, 13 | 2661 | |||
GenBank mRNA | AB208934.1 | - | 11, 12 | 1315 | ||
AF030339.1 | - | 12, 13 | 2634 |