Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168679.1 | NP_001162150.1 | 7, 8 | 755 | ||
NM_001168680.1 | NP_001162151.1 | 7, 8 | 791 | |||
NM_146010.2 | NP_666122.1 | 6, 7 | 897 | |||
GenBank mRNA | AK077301.1 | - | 7, 8 | 805 | ||
AK087696.1 | - | 7, 8 | 831 | |||
BC025461.1 | - | 7, 8 | 763 | 5344986 | ||
BC027777.1 | - | 7, 8 | 766 | 5351814 | ||
BC029068.1 | - | 7, 8 | 787 | 4947664 | ||
BC029808.1 | - | 6, 7 | 896 | 4947699 | ||
BC034198.1 | - | 6, 7 | 670 | 4986549 | ||
DV060937.1 | - | 6, 7 | 730 |