Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005783.3 | NP_005774.2 | 5 | 845 | ||
GenBank mRNA | AB006679.1 | - | 5 | 783 | ||
AB451403.1 | - | 4 | 654 | |||
AK222616.1 | - | 5 | 771 | |||
AK313777.1 | - | 5 | 765 | |||
BC005968.1 | - | 5 | 781 | 4073821 | ||
BC022864.1 | - | 5 | 785 | 4096526 | ||
BC070183.1 | - | 5 | 779 | 4551680 | ||
BM553565.1 | - | 161 | ||||
CR456935.1 | - | 4 | 654 |