Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177669.1 | NP_001171140.1 | 4 | 1048 | ||
NM_146133.3 | NP_666245.2 | 5 | 1237 | |||
GenBank mRNA | AK008434.1 | - | 5 | 1204 | ||
AK160454.1 | - | 4 | 1009 | |||
BC027194.1 | - | 5 | 1147 | 4216692 | ||
BC047147.1 | - | 5 | 1212 | 6465983 |