Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146150.2 | NP_666262.2 | 31 | 3545 | ||
GenBank mRNA | AK031548.1 | - | 31 | 3545 | ||
AK050944.1 | - | 16 | 1644 |