Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153057.4 | NP_694697.3 | 28 | 3348 | ||
GenBank mRNA | AK030584.1 | - | 28 | 3316 | ||
AK035957.1 | - | 28 | 3320 | |||
AK043279.1 | - | 3 | 165 | |||
AK080754.1 | - | 28 | 3211 | |||
AK082495.1 | - | 28 | 3227 | |||
AK082963.1 | - | 28 | 3207 | |||
AK142338.1 | - | 28 | 3318 | |||
AK165643.1 | - | 28 | 3212 | |||
AK166764.1 | - | 13 | 1358 | |||
AK190977.1 | - | 2 | 51 | |||
AK200117.1 | - | 2 | 51 | |||
AK216429.1 | - | 2 | 51 | |||
BC022138.1 | - | 27 | 3064 | |||
BC024503.1 | - | 6 | 591 | |||
BC025133.1 | - | 11 | 1227 | |||
BC033923.1 | - | 17 | 1932 | 5099899 | ||
BC072630.1 | - | 28 | 3294 | 6833516 |