Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153088.2 | NP_694728.1 | 3, 4 | 618 | ||
GenBank mRNA | AK089478.1 | - | 3, 4 | 526 | ||
AK167390.1 | - | 3, 4 | 376 | |||
AY028804.1 | - | 3, 4 | 643 | |||
BC049184.1 | - | 3, 4 | 478 | 6439402 | ||
BC058085.1 | - | 2, 3 | 805 | |||
BC065158.1 | - | 3, 4 | 353 | 5717102 | ||
BC079638.1 | - | 3, 4 | 353 | 5717102 |