Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145948.1 | NP_001139420.1 | 4 | 370 | ||
NM_153392.2 | NP_700441.2 | 4 | 379 | |||
GenBank mRNA | AK048659.1 | - | 4 | 380 | ||
AK076641.1 | - | 4 | 332 | |||
AK076720.1 | - | 4 | 371 | |||
AK076772.1 | - | 4 | 333 | |||
AK082733.1 | - | 4 | 370 | |||
AK172951.1 | - | 4 | 302 | |||
BC023883.1 | - | 4 | 378 | 5150841 | ||
BC026660.1 | - | 4 | 320 | 4988900 |