Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145948.1 | NP_001139420.1 | 6, 7 | 541 | ||
NM_153392.2 | NP_700441.2 | 6, 7 | 550 | |||
GenBank mRNA | AK048659.1 | - | 6, 7 | 551 | ||
AK076641.1 | - | 6, 7 | 503 | |||
AK076720.1 | - | 6, 7 | 542 | |||
AK076772.1 | - | 6, 7 | 504 | |||
AK172951.1 | - | 6, 7 | 473 | |||
BC023883.1 | - | 6, 7 | 549 | 5150841 | ||
BC026660.1 | - | 6, 7 | 491 | 4988900 |