Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153546.4 | NP_705774.1 | 7 | 794 | ||
GenBank mRNA | AB297382.1 | - | 7 | 617 | ||
AB305045.1 | - | 7 | 617 | |||
AK033388.1 | - | 7 | 736 | |||
AK137755.1 | - | 7 | 726 | |||
AK146984.1 | - | 7 | 701 | |||
BC023845.1 | - | 7 | 798 | 5346000 | ||
BC024653.1 | - | 7 | 746 | 3660479 |