Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153783.4 | NP_722478.2 | 4 | 913 | ||
GenBank mRNA | AF226656.2 | - | 4 | 932 | ||
AK030664.1 | - | 4 | 914 | |||
AK159137.1 | - | 2 | 200 | |||
AK186235.1 | - | 2 | 67 | |||
AM049402.1 | - | 4 | 905 | |||
BC033913.1 | - | 3 | 177 | |||
BC082783.1 | - | 4 | 912 | 6475809 |