Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153790.2 | NP_722485.1 | 6, 7 | 1279 | ||
GenBank mRNA | AB024432.1 | - | 6, 7 | 1246 | ||
AF522197.1 | - | 6, 7 | 1280 | |||
AK142378.1 | - | 6, 7 | 1247 | |||
AK143208.1 | - | 6, 7 | 1122 | |||
AK170331.1 | - | 2, 3 | 333 | |||
BC059226.1 | - | 6, 7 | 1211 | |||
BC060084.1 | - | 6, 7 | 1211 | |||
BC139758.1 | - | 6, 7 | 1232 | 40047931 |