Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134829.1 | NP_001128301.1 | 6 | 980 | ||
NM_172266.3 | NP_758470.1 | 6 | 959 | |||
GenBank mRNA | AK133814.1 | - | 6 | 960 | ||
AK172914.1 | - | 6 | 930 | |||
BC013667.1 | - | 6 | 868 | 4020003 | ||
BC024750.1 | - | 4 | 450 |