Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172501.2 | NP_766089.1 | 6 | 816 | ||
GenBank mRNA | AK029465.1 | - | 6 | 811 | ||
AK033167.1 | - | 6 | 816 | |||
AK133986.1 | - | 766 | ||||
AK220477.1 | - | 733 | ||||
BC087903.1 | - | 6 | 803 | 30250042 | ||
BC107356.1 | - | 6 | 804 | 40055875 |