Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166669.1 | NP_001160141.1 | 4, 5 | 745 | ||
NM_001166671.1 | NP_001160143.1 | 4, 5 | 624 | |||
NM_172558.3 | NP_766146.2 | 4, 5 | 745 | |||
GenBank mRNA | AK049216.1 | - | 4, 5 | 680 | ||
AK131137.1 | - | 4, 5 | 600 | |||
AK143536.1 | - | 4, 5 | 694 | |||
BC137802.1 | - | 4, 5 | 688 | 8860821 | ||
BC144922.1 | - | 4, 5 | 688 | 9053470 |