Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172568.2 | NP_766156.2 | 8, 9 | 2416 | ||
GenBank mRNA | AK077168.1 | - | 2, 3 | 1036 | ||
BC117499.1 | - | 2, 3 | 1030 |