Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172584.3 | NP_766172.1 | 10 | 2558 | ||
GenBank mRNA | AK038913.1 | - | 10 | 2560 | ||
AK185559.1 | - | 1 | 158 | |||
AK187570.1 | - | 1 | 257 | |||
BC025917.1 | - | 3 | 1838 | |||
BC031182.1 | - | 2 | 1620 | |||
BC056464.1 | - | 10 | 2539 | 5707377 |