Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172599.4 | NP_766187.1 | 3 | 345 | ||
GenBank mRNA | AK029330.1 | - | 3 | 326 | ||
AK029967.1 | - | 3 | 326 | |||
AK054196.1 | - | 3 | 346 | |||
BC057360.1 | - | 2 | 65 | |||
BC090995.1 | - | 3 | 325 | 6754513 |