Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177627.1 | NP_001171098.1 | 7 | 1031 | ||
NM_172659.2 | NP_766247.2 | 7 | 1031 | |||
GenBank mRNA | AK079650.1 | - | 7 | 975 | ||
AK089246.1 | - | 7 | 1023 | |||
AK137441.1 | - | 6 | 1246 | |||
AK150104.1 | - | 7 | 984 | |||
AK163445.1 | - | 7 | 989 | |||
AK166312.1 | - | 7 | 989 | |||
AK172411.1 | - | 7 | 970 | |||
BC025498.1 | - | 4 | 1615 | |||
BC058210.1 | - | 7 | 955 | |||
BC141168.1 | - | 7 | 962 | 9055780 | ||
BC145480.1 | - | 7 | 957 |