Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080754.1 | NP_001074223.1 | 5 | 819 | ||
NM_172669.3 | NP_766257.3 | 5 | 819 | |||
GenBank mRNA | AK037516.1 | - | 5 | 776 | ||
AK083596.1 | - | 5 | 776 | |||
AK084557.1 | - | 5 | 800 | |||
AK129433.1 | - | 1 | 83 | |||
AK148887.1 | - | 5 | 811 | |||
AK155513.1 | - | 5 | 782 | |||
AK169280.1 | - | 5 | 772 | |||
BC032293.1 | - | 5 | 742 | |||
BC054082.1 | - | 5 | 751 |