Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172687.1 | NP_766275.1 | 7 | 1530 | ||
GenBank mRNA | AK028680.1 | - | 7 | 1530 | ||
AK035318.1 | - | 7 | 1525 | |||
AK142287.1 | - | 7 | 1468 | |||
BC016572.1 | - | 1 | 279 |