Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205363.1 | NP_001192292.1 | 7 | 3115 | ||
NM_001205364.1 | NP_001192293.1 | 6 | 3034 | |||
NM_001205365.1 | NP_001192294.1 | 4 | 2766 | |||
NM_172732.3 | NP_766320.2 | 6 | 3037 | |||
GenBank mRNA | AK036399.1 | - | 7 | 3117 |