Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prune | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173347.2 | NP_775482.1 | 3, 4 | 782 | ||
GenBank mRNA | AF051908.2 | - | 3, 4 | 329 | ||
AK081637.1 | - | 3, 4 | 473 | |||
AK142160.1 | - | 3, 4 | 381 | |||
BC044877.1 | - | 1, 2 | 207 | |||
BC057546.1 | - | 3, 4 | 782 | 6416282 | ||
BC058635.1 | - | 3, 4 | 511 | 5698271 |