Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162375.1 | NP_001155847.1 | 9 | 1348 | ||
NM_174868.4 | NP_777357.2 | 11 | 1540 | |||
GenBank mRNA | AK044739.1 | - | 11 | 1516 | ||
AK045347.1 | - | 9 | 1347 | |||
AK083909.1 | - | 10 | 1371 | |||
AK085017.1 | - | 11 | 1540 | |||
AK143366.1 | - | 11 | 1540 | |||
AK166051.1 | - | 11 | 1493 | |||
BC098205.1 | - | 11 | 1523 | 6409928 | ||
BC138328.1 | - | 11 | 1487 | 8861348 | ||
BC138329.1 | - | 11 | 1487 | 8861349 |