Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297612.1 | NP_001284541.1 | 3 | 289 | ||
NM_175441.5 | NP_780650.2 | 3 | 742 | |||
GenBank mRNA | AK052858.1 | - | 3 | 577 | ||
AK146683.1 | - | 3 | 743 | |||
AK165216.1 | - | 3 | 228 | |||
BB843262.1 | - | 3 | 290 | |||
BC137983.1 | - | 3 | 715 | 8861002 | ||
BC145079.1 | - | 3 | 715 | |||
EU403565.1 | - | 3 | 685 |