Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297612.1 | NP_001284541.1 | 4 | 922 | ||
NM_175441.5 | NP_780650.2 | 4 | 1375 | |||
GenBank mRNA | AK052858.1 | - | 4 | 1210 | ||
AK146683.1 | - | 4 | 1376 | |||
AK165216.1 | - | 4 | 861 | |||
BC137983.1 | - | 4 | 1348 | 8861002 | ||
BC145079.1 | - | 4 | 1349 | |||
EU403565.1 | - | 4 | 1318 |