Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175937.3 | NP_787951.2 | 3 | 1983 | ||
GenBank mRNA | AB100307.1 | - | 4 | 616 | ||
BC107349.1 | - | 4 | 593 | 40055453 | ||
BC107350.1 | - | 4 | 593 | 40055458 |