Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178149.4 | NP_835362.2 | 6 | 1708 | ||
GenBank mRNA | AK154842.1 | - | 6 | 1650 | ||
AK161428.1 | - | 2 | 1066 | |||
AY522649.1 | - | 6 | 2038 | |||
BC053972.1 | - | 1 | 396 | |||
BC055920.1 | - | 6 | 1645 | 4039129 | ||
BC079641.1 | - | 6 | 1632 | 5707778 |