Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178241.4 | NP_839972.1 | 2 | 599 | ||
GenBank mRNA | AY227797.1 | - | 1 | 551 | ||
AY390263.1 | - | 2 | 599 | |||
AY749637.1 | - | 1 | 925 |