Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105562.2 | NP_001099032.1 | 3 | 937 | ||
NM_006048.4 | NP_006039.2 | 3 | 937 | |||
XM_005263422.1 | XP_005263479.1 | 3 | 761 | |||
XM_011540489.2 | XP_011538791.1 | 3 | 761 | |||
XM_011540490.1 | XP_011538792.1 | 3 | 761 | |||
XM_017000036.1 | XP_016855525.1 | 3 | 524 | |||
GenBank mRNA | AB014584.3 | - | 3 | 828 | ||
AB028839.1 | - | 3 | 945 | |||
AF043117.1 | - | 3 | 334 | |||
AF331520.1 | - | 3 | 296 | |||
AK292444.1 | - | 3 | 419 | |||
BC093696.1 | - | 3 | 327 | 7939541 | ||
BC143458.1 | - | 3 | 327 | |||
BC143460.1 | - | 3 | 327 | |||
JF289274.1 | - | 3 | 249 |